• USA Home
  • 8.41083 - 2,4-Dichlorobenzyl alcohol

EMAIL THIS PAGE TO A FRIEND
8.41083 EMD Millipore

2,4-Dichlorobenzyl alcohol

2,4-Dichlorobenzyl alcohol for synthesis. CAS 1777-82-8, molar mass 177.03 g/mol., for synthesis

Synonym: Dybenal

  • CAS Number 1777-82-8

  • Linear Formula C₇H₆Cl₂O

  • Molecular Weight 177.03 g/mol

  •  EC Number 217-210-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (8410830025)
solubility   1 g/l
mp   56 - 58 °C
bulk density   1450 kg/m3
LD 50 oral   LD50 Rat > 2000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 58 °C
  Melting range (upper value): ≤ 60 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2906 29 00
Hazard Statement(s) 
H318: Causes serious eye damage.
Precautionary Statement(s) 
P280: Wear eye protection.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P313: Get medical advice/ attention.
RTECS 
DO0890000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Symbols 
Irritant  Irritant
Categories of danger 
irritant
R Phrase 
R 41
Risk of serious damage to eyes.
S Phrase 
S 26-39
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear eye/face protection.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?