• USA Home
  • 8.41090 - 4-Nitrobenzophenone

EMAIL THIS PAGE TO A FRIEND
8.41090 EMD Millipore

4-Nitrobenzophenone

4-Nitrobenzophenone for synthesis. CAS 1144-74-7, molar mass 227.22 g/mol., for synthesis

Synonym: (4-Nitrophenyl)phenylmethanone

  • CAS Number 1144-74-7

  • Linear Formula C₁₃H₉NO₃

  • Molecular Weight 227.22 g/mol

  •  EC Number 214-542-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Glass bottle (8410900010)
mp   136 - 137 °C
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 136 °C
  Melting range (upper value): ≤ 138 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2914 79 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?