• USA Home
  • 8.41093 - 2-Ethyl-6-methylaniline

EMAIL THIS PAGE TO A FRIEND
8.41093 EMD Millipore

2-Ethyl-6-methylaniline

2-Ethyl-6-methylaniline for synthesis. CAS 24549-06-2, molar mass 135.2 g/mol., for synthesis

Synonym: 6-Ethyl-o-toluidine

  • CAS Number 24549-06-2

  • Linear Formula C₉H₁₃N

  • Molecular Weight 135.2 g/mol

  •  EC Number 246-309-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8410930100)
bp   231 °C (1013 hPa)
vapor pressure   0.07 hPa (20 °C)
solubility   2.66 g/l
density   0.969 g/cm3 (20 °C)
flash point   105 °C
autoignition temp.   460 °C
mp   -24 °C
LD 50 oral   LD50 Rat 1150 mg/kg
LD 50 dermal   LD50 Rat 1290 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.966 - 0.970
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2921 49 00
Hazard Statement(s) 
H302 + H312 + H332: Harmful if swallowed, in contact with skin or if inhaled
H412: Harmful to aquatic life with long lasting effects.
Precautionary Statement(s) 
P262: Do not get in eyes, on skin, or on clothing.
P273: Avoid release to the environment.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 2810 , 6.1, III
Declaration (transport by air) IATA-DGR 
UN 2810 , 6.1, III
Declaration (transport by sea) IMDG-Code 
UN 2810 , 6.1, III
Categories of danger 
harmful, dangerous for the environment
R Phrase 
R 20/21/22-52/53
Harmful by inhalation, in contact with skin and if swallowed.Harmful to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
S Phrase 
S 25-36/37-61
Avoid contact with eyes.Wear suitable protective clothing and gloves.Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?