• USA Home
  • 8.41095 - 2,6-Diisopropylaniline

EMAIL THIS PAGE TO A FRIEND
8.41095 EMD Millipore

2,6-Diisopropylaniline

2,6-Diisopropylaniline for synthesis. CAS 24544-04-5, molar mass 177.29 g/mol., for synthesis

Synonym: 2,6-Bis(isopropyl)-benzenamine

  • CAS Number 24544-04-5

  • Linear Formula C₁₂H₁₉N

  • Molecular Weight 177.29 g/mol

  •  EC Number 246-305-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
bp   257 °C (1013 hPa)
solubility   <0.2 g/l
density   0.94 g/cm3 (20 °C)
flash point   125 °C
autoignition temp.   400 °C DIN 51794
mp   -45 °C
LD 50 oral   LD50 Rat 4077 mg/kg
LD 50 dermal   LD50 Rat > 2000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 92.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.937 - 0.941
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2921 49 00
Hazard Statement(s) 
H319: Causes serious eye irritation.
H412: Harmful to aquatic life with long lasting effects.
Precautionary Statement(s) 
P273: Avoid release to the environment.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RTECS 
BX4025000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Categories of danger 
irritant, dangerous for the environment
R Phrase 
R 36-52/53
Irritating to eyes.Harmful to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
S Phrase 
S 61
Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?