• USA Home
  • 8.41096 - tert-Butyl cyanoacetate

EMAIL THIS PAGE TO A FRIEND
8.41096 EMD Millipore

tert-Butyl cyanoacetate

tert-Butyl cyanoacetate for synthesis. CAS 1116-98-9, molar mass 141.16 g/mol., for synthesis

Synonym: Cyanoacetic acid tert-butyl ester

  • CAS Number 1116-98-9

  • Linear Formula C₇H₁₁NO₂

  • Molecular Weight 141.16 g/mol

  •  EC Number 214-243-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8410960025)
bp   67 - 68 °C (2 hPa)
density   0.987 g/cm3 (20 °C)
flash point   91 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.985 - 0.989
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2926 90 70
Storage class 
6.1C Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Declaration (railroad and road) ADR, RID 
Kein Gefahrgut
Declaration (transport by air) IATA-DGR 
No Dangerous Good
Declaration (transport by sea) IMDG-Code 
No Dangerous Good
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?