• USA Home
  • 8.41098 - 4-Methylcinnamic acid

EMAIL THIS PAGE TO A FRIEND
8.41098 EMD Millipore

4-Methylcinnamic acid

4-Methylcinnamic acid for synthesis. CAS 1866-39-3, molar mass 162.19 g/mol., for synthesis

Synonym: 3-(4-Tolyl)acrylic acid

  • CAS Number 1866-39-3

  • Linear Formula C₁₀H₁₀O₂

  • Molecular Weight 162.19 g/mol

  •  EC Number 217-479-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Glass bottle (8410980010)
mp   196 - 198 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 196 °C
  Melting range (upper value): ≤ 200 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2916 39 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?