EMAIL THIS PAGE TO A FRIEND
8.41110 EMD Millipore

5-Hydroxyindole

5-Hydroxyindole for synthesis. CAS 1953-54-4, molar mass 133.15 g/mol., for synthesis

Synonym: 5-Indolol

  • CAS Number 1953-54-4

  • Linear Formula C₈H₇NO

  • Molecular Weight 133.15 g/mol

  •  EC Number 217-782-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   1 g in Glass bottle (8411100001)
solubility   38 g/l
mp   107 - 108 °C
storage temp.   Store at <= 20°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 105 °C
  Melting range (upper value): ≤ 108 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 99 80
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?