• USA Home
  • 8.41114 - 2-Methyl-5-nitrobenzoic acid

EMAIL THIS PAGE TO A FRIEND
8.41114 EMD Millipore

2-Methyl-5-nitrobenzoic acid

2-Methyl-5-nitrobenzoic acid for synthesis. CAS 1975-52-6, molar mass 181.15 g/mol., for synthesis

Synonym: 5-Nitro-o-toluic acid

  • CAS Number 1975-52-6

  • Linear Formula C₈H₇NO₄

  • Molecular Weight 181.15 g/mol

  •  EC Number 217-829-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Glass bottle (8411140005)
mp   176 - 180 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (acidimetric): ≥ 98.5 %
  Melting range (lower value): ≥ 176 °C
  Melting range (upper value): ≤ 180 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2916 39 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?