• USA Home
  • 8.41115 - Bis(diphenylphosphino)-methane

EMAIL THIS PAGE TO A FRIEND
8.41115 EMD Millipore

Bis(diphenylphosphino)-methane

Bis(diphenylphosphino)-methane for synthesis. CAS 2071-20-7, molar mass 384.39 g/mol., for synthesis

Synonym: Methylenebis(diphenylphosphine)

  • CAS Number 2071-20-7

  • Linear Formula C₂₅H₂₂P₂

  • Molecular Weight 384.39 g/mol

  •  EC Number 218-194-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Plastic bottle (8411150005)
mp   118 - 119 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 96.0 % (a/a)
  Melting range (lower value): ≥ 118 °C
  Melting range (upper value): ≤ 121 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2931 39 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?