• USA Home
  • 8.41117 - 2,6-Difluorobenzonitrile

EMAIL THIS PAGE TO A FRIEND
8.41117 EMD Millipore

2,6-Difluorobenzonitrile

2,6-Difluorobenzonitrile for synthesis. CAS 1897-52-5, molar mass 139.1 g/mol., for synthesis

  • CAS Number 1897-52-5

  • Linear Formula C₇H₃F₂N

  • Molecular Weight 139.1 g/mol

  •  EC Number 217-589-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 ml in Glass bottle (8411170010)
density   1.246 g/cm3 (20 °C)
flash point   90 °C
mp   30 - 32 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2926 90 70
Hazard Statement(s) 
H302 + H312: Harmful if swallowed or in contact with skin
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 3439 , 6.1, III
Declaration (transport by air) IATA-DGR 
UN 3439 , 6.1, III
Declaration (transport by sea) IMDG-Code 
UN 3439 , 6.1, III
Categories of danger 
harmful
R Phrase 
R 21/22
Harmful in contact with skin and if swallowed.
S Phrase 
S 36/37
Wear suitable protective clothing and gloves.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?