• USA Home
  • 8.41118 - Diethyl allylmalonate

EMAIL THIS PAGE TO A FRIEND
8.41118 EMD Millipore

Diethyl allylmalonate

Diethyl allylmalonate for synthesis. CAS 2049-80-1, molar mass 200.23 g/mol., for synthesis

Synonym: 2-Propenyl-propanedioic acid diethyl ester, Allylmalonic acid diethyl ester

  • CAS Number 2049-80-1

  • Linear Formula C₁₀H₁₆O₄

  • Molecular Weight 200.23 g/mol

  •  EC Number 218-072-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8411180025)
bp   105 - 107 °C (22.7 hPa)
density   1.016 g/cm3 (20 °C)
flash point   92 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 95.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.000 - 1.010
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2917 19 80
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?