• USA Home
  • 8.41125 - Thiophene-2-acetonitrile

EMAIL THIS PAGE TO A FRIEND
8.41125 EMD Millipore

Thiophene-2-acetonitrile

Thiophene-2-acetonitrile for synthesis. CAS 20893-30-5, EC Number 244-104-6, molar mass 123.17 g/mol., for synthesis

Synonym: 2-Cyanomethylthiophene

  • CAS Number 20893-30-5

  • Linear Formula C₆H₅NS

  • Molecular Weight 123.17 g/mol

  •  EC Number 244-104-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8411250025)
bp   110 - 114 °C (18.7 hPa)
density   1.16 g/cm3 (20 °C)
flash point   101 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 96.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.156 - 1.160
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2934 99 90
Storage class 
6.1C Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Declaration (railroad and road) ADR, RID 
UN 2810 , 6.1, III
Declaration (transport by air) IATA-DGR 
UN 2810 , 6.1, III
Declaration (transport by sea) IMDG-Code 
UN 2810 , 6.1, III
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?