• USA Home
  • 8.41126 - 2-Bromobenzyl alcohol

EMAIL THIS PAGE TO A FRIEND
8.41126 EMD Millipore

2-Bromobenzyl alcohol

2-Bromobenzyl alcohol for synthesis. CAS 18982-54-2, molar mass 187.04 g/mol., for synthesis

  • CAS Number 18982-54-2

  • Linear Formula C₇H₇BrO

  • Molecular Weight 187.04 g/mol

  •  EC Number 242-719-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Plastic bottle (8411260010)
mp   80 - 82 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 78 °C
  Melting range (upper value): ≤ 81 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2906 29 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?