• USA Home
  • 8.41127 - 3-(3,4-Dimethoxyphenyl)-propionic acid

EMAIL THIS PAGE TO A FRIEND
8.41127 EMD Millipore

3-(3,4-Dimethoxyphenyl)-propionic acid

3-(3,4-Dimethoxyphenyl)-propionic acid for synthesis. CAS 2107-70-2, molar mass 210.22 g/mol., for synthesis

Synonym: 3,4-Dimethoxyhydrocinnamic acid

  • CAS Number 2107-70-2

  • Linear Formula C₁₁H₁₄O₄

  • Molecular Weight 210.22 g/mol

  •  EC Number 218-288-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   50 g in Glass bottle (8411270050)
mp   99 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (acidimetric): ≥ 98.0 %
  Melting range (lower value): ≥ 97 °C
  Melting range (upper value): ≤ 99 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2918 99 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?