• USA Home
  • 8.41130 - 3-Bromophenylacetic acid

EMAIL THIS PAGE TO A FRIEND
8.41130 EMD Millipore

3-Bromophenylacetic acid

3-Bromophenylacetic acid for synthesis. CAS 1878-67-7, molar mass 215.04 g/mol., for synthesis

  • CAS Number 1878-67-7

  • Linear Formula C₈H₇BrO₂

  • Molecular Weight 215.04 g/mol

  •  EC Number 217-522-1

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   1 g in Plastic bottle (8411300001)
mp   102 - 103 °C
storage temp.   Store below +30°C.
specifications   Assay (acidimetric): ≥ 98.0 %
  Melting range (lower value): ≥ 100 °C
  Melting range (upper value): ≤ 103 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2916 39 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?