EMAIL THIS PAGE TO A FRIEND
8.41136 EMD Millipore

5-Chlorouracil

5-Chlorouracil for synthesis. CAS 1820-81-1, molar mass 146.53 g/mol., for synthesis

Synonym: 5-Chloro-2,4-dihydroxypyrimidine

  • CAS Number 1820-81-1

  • Linear Formula C₄H₃ClN₂O₂

  • Molecular Weight 146.53 g/mol

  •  EC Number 217-339-7

Purchase

Properties

Related Categories Biochemicals and Reagents, Building Blocks, Carbonyl Compounds, Chemical Synthesis, Organic Building Blocks,
packaging   5 g in Plastic bottle (8411360005)
mp   >300 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 99.0 % (a/a)
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 59 95
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?