• USA Home
  • 8.41140 - 3-Bromobenzyl alcohol

EMAIL THIS PAGE TO A FRIEND
8.41140 EMD Millipore

3-Bromobenzyl alcohol

3-Bromobenzyl alcohol for synthesis. CAS 15852-73-0, molar mass 187.03 g/mol., for synthesis

  • CAS Number 15852-73-0

  • Linear Formula C₇H₇BrO

  • Molecular Weight 187.03 g/mol

  •  EC Number 239-975-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 ml in Glass bottle (8411400005)
bp   165 °C (21.3 hPa)
density   1.56 g/cm3 (20 °C)
flash point   >110 °C
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.550 - 1.558
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2906 29 00
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?