• USA Home
  • 8.41144 - 3,5-Bis(trifluoromethyl)-aniline

EMAIL THIS PAGE TO A FRIEND
8.41144 EMD Millipore

3,5-Bis(trifluoromethyl)-aniline

3,5-Bis(trifluoromethyl)-aniline for synthesis. CAS 328-74-5, molar mass 229.12 g/mol., for synthesis

Synonym: α,α,α,α',α',α'-Hexafluoro-3,5-xylidine

  • CAS Number 328-74-5

  • Linear Formula C₈H₅F₆N

  • Molecular Weight 229.12 g/mol

  •  EC Number 206-335-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 ml in Glass bottle (8411440010)
bp   75 - 78 °C (13 hPa)
density   1.48 g/cm3 (20 °C)
flash point   83 °C
mp   3 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.478 - 1.483
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2921 49 00
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Declaration (railroad and road) ADR, RID 
UN 2810 , 6.1, II
Declaration (transport by air) IATA-DGR 
UN 2810 , 6.1, II
Declaration (transport by sea) IMDG-Code 
UN 2810 , 6.1, II
Categories of danger 
irritant
R Phrase 
R 36/38
Irritating to eyes and skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?