• USA Home
  • 8.41145 - N-tert-BOC-Hydroxylamine

EMAIL THIS PAGE TO A FRIEND
8.41145 EMD Millipore

N-tert-BOC-Hydroxylamine

N-tert-BOC-Hydroxylamine for synthesis. CAS 36016-38-3, molar mass 133.15 g/mol., for synthesis

Synonym: tert-Butyl N-hydroxycarbamate

  • CAS Number 36016-38-3

  • Linear Formula C₅H₁₁NO₃

  • Molecular Weight 133.15 g/mol

  •  EC Number 252-836-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Glass bottle (8411450005)
mp   55 °C
shipped in   ambient
storage temp.   Store at <= 20°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 55 °C
  Melting range (upper value): ≤ 57 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2928 00 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?