• USA Home
  • 8.41146 - 4-Biphenylyl-methanol

EMAIL THIS PAGE TO A FRIEND
8.41146 EMD Millipore

4-Biphenylyl-methanol

4-Biphenylyl-methanol for synthesis. CAS 3597-91-9, EC Number 222-745-2, molar mass 184.23 g/mol., for synthesis

Synonym: p-Phenylbenzyl alcohol, 4-Hydroxymethylbiphenyl

  • CAS Number 3597-91-9

  • Linear Formula C₁₃H₁₂O

  • Molecular Weight 184.23 g/mol

  •  EC Number 222-745-2

Purchase

Properties

packaging   10 g in Plastic bottle (8411460010)
mp   104 - 105 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 99 °C
  Melting range (upper value): ≤ 104 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2906 29 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?