• USA Home
  • 8.41149 - Heptafluoro-1-butanol

EMAIL THIS PAGE TO A FRIEND
8.41149 EMD Millipore

Heptafluoro-1-butanol

Heptafluoro-1-butanol for synthesis. CAS 375-01-9, molar mass 200.06 g/mol., for synthesis

  • CAS Number 375-01-9

  • Linear Formula C₄H₃F₇O

  • Molecular Weight 200.06 g/mol

  •  EC Number 206-782-1

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 ml in Glass bottle (8411490010)
density   1.596 g/cm3 (20 °C)
flash point   25 °C
LD 50 oral   LD50 Rat 3630 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2905 59 98
Hazard Statement(s) 
H226: Flammable liquid and vapour.
H319: Causes serious eye irritation.
H315: Causes skin irritation.
Precautionary Statement(s) 
P210: Keep away from heat.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
RTECS 
EL4130000
Storage class 
3 Flammable liquids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Declaration (railroad and road) ADR, RID 
UN 1993 , 3, II
Declaration (transport by air) IATA-DGR 
UN 1993 , 3, II
Declaration (transport by sea) IMDG-Code 
UN 1993 , 3, II
Categories of danger 
flammable, irritant
R Phrase 
R 10-36/38
Flammable.Irritating to eyes and skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?