• USA Home
  • 8.41153 - 3-Methoxy-2-nitrobenzoic acid

EMAIL THIS PAGE TO A FRIEND
8.41153 EMD Millipore

3-Methoxy-2-nitrobenzoic acid

3-Methoxy-2-nitrobenzoic acid for synthesis. CAS 4920-80-3, molar mass 197.15 g/mol., for synthesis

Synonym: 2-Nitro-m-anisic acid

  • CAS Number 4920-80-3

  • Linear Formula C₈H₇NO₅

  • Molecular Weight 197.15 g/mol

  •  EC Number 225-549-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
mp   259 - 262 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (acidimetric): ≥ 98.0 %
  Melting range (lower value): ≥ 256 °C
  Melting range (upper value): ≤ 260 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2918 99 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?