• USA Home
  • 8.41157 - 3'-(Trifluoromethyl)-acetophenone

EMAIL THIS PAGE TO A FRIEND
8.41157 EMD Millipore

3'-(Trifluoromethyl)-acetophenone

3'-(Trifluoromethyl)-acetophenone for synthesis. CAS 349-76-8, molar mass 188.14 g/mol., for synthesis

Synonym: 3-Acetylbenzotrifluoride

  • CAS Number 349-76-8

  • Linear Formula C₉H₇F₃O

  • Molecular Weight 188.14 g/mol

  •  EC Number 206-490-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 ml in Glass bottle (8411570005)
density   1.26 g/cm3 (20 °C)
flash point   83 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.260 - 1.266
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2914 79 00
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
H335: May cause respiratory irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Categories of danger 
irritant
R Phrase 
R 36/37/38
Irritating to eyes, respiratory system and skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?