• USA Home
  • 8.41171 - Ethyl 4-fluorobenzoate

EMAIL THIS PAGE TO A FRIEND
8.41171 EMD Millipore

Ethyl 4-fluorobenzoate

Ethyl 4-fluorobenzoate for synthesis. CAS 451-46-7, molar mass 168.16 g/mol., for synthesis

Synonym: 4-Fluorobenzoic acid ethyl ester

  • CAS Number 451-46-7

  • Linear Formula C₉H₉FO₂

  • Molecular Weight 168.16 g/mol

  •  EC Number 207-194-8

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 ml in Glass bottle (8411710010)
bp   209 - 210 °C
density   1.146 g/cm3 (20 °C)
flash point   81 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 22 °C
  Melting range (upper value): ≤ 25 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2916 39 90
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?