• USA Home
  • 8.41178 - 2-Furancarboxylic acid hydrazide

EMAIL THIS PAGE TO A FRIEND
8.41178 EMD Millipore

2-Furancarboxylic acid hydrazide

2-Furancarboxylic acid hydrazide for synthesis. CAS 3326-71-4, molar mass 126.11 g/mol., for synthesis

Synonym: 2-Furoic acid hydrazide

  • CAS Number 3326-71-4

  • Linear Formula C₅H₆N₂O₂

  • Molecular Weight 126.11 g/mol

  •  EC Number 222-046-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Plastic bottle (8411780010)
mp   79 - 81 °C
shipped in   ambient
storage temp.   Store at +15°C to +25°C.
specifications   Assay (GC, area%): ≥ 96.0 % (a/a)
  Melting range (lower value): ≥ 73 °C
  Melting range (upper value): ≤ 80 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2932 19 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?