• USA Home
  • 8.41181 - Ethyl heptafluorobutyrate

EMAIL THIS PAGE TO A FRIEND
8.41181 EMD Millipore

Ethyl heptafluorobutyrate

Ethyl heptafluorobutyrate for synthesis. CAS 356-27-4, molar mass 242.09 g/mol., for synthesis

Synonym: Heptafluoro-butanoic acid ethyl ester

  • CAS Number 356-27-4

  • Linear Formula C₆H₅F₇O₂

  • Molecular Weight 242.09 g/mol

  •  EC Number 206-602-1

Purchase

Properties

Related Categories Biochemicals and Reagents More...
bp   95 - 98 °C
density   1.398 g/cm3 (20 °C)
flash point   14 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 96.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.395 - 1.400
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2915 90 70
Hazard Statement(s) 
H225: Highly flammable liquid and vapour.
Precautionary Statement(s) 
P210: Keep away from heat/sparks/open flames/hot surfaces. No smoking.
P262: Do not get in eyes, on skin, or on clothing.
P403 + P235: Store in a well-ventilated place. Keep cool.
Storage class 
3 Flammable liquids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Symbols 
Flammable  Flammable
Declaration (railroad and road) ADR, RID 
UN 3272 , 3, II
Declaration (transport by air) IATA-DGR 
UN 3272 , 3, II
Declaration (transport by sea) IMDG-Code 
UN 3272 , 3, II
Categories of danger 
highly flammable
R Phrase 
R 11
Highly flammable.
S Phrase 
S 24/25
Avoid contact with skin and eyes.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?