• USA Home
  • 8.41185 - N-(2-Hydroxyethyl)-phthalimide

EMAIL THIS PAGE TO A FRIEND
8.41185 EMD Millipore

N-(2-Hydroxyethyl)-phthalimide

for synthesis

Synonym: 2-Phthalimidoethanol

  • CAS Number 3891-07-4

  • Linear Formula C₁₀H₉NO₃

  • Molecular Weight 191.19 g/mol

  •  EC Number 223-434-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Plastic bottle (8411850100)
mp   127 - 130 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 127 °C
  Melting range (upper value): ≤ 130 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2925 19 95
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?