• USA Home
  • 8.41190 - 4-(Methylthio)benzaldehyde

EMAIL THIS PAGE TO A FRIEND
8.41190 EMD Millipore

4-(Methylthio)benzaldehyde

4-(Methylthio)benzaldehyde for synthesis. CAS 3446-89-7, molar mass 152.22 g/mol., for synthesis

Synonym: 4-(Methylmercapto)benzaldehyde

  • CAS Number 3446-89-7

  • Linear Formula C₈H₈OS

  • Molecular Weight 152.22 g/mol

  •  EC Number 222-365-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8411900025)
bp   118 - 121 °C (16 hPa)
density   1.144 g/cm3 (20 °C)
flash point   >110 °C
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.166 - 1.171
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2930 90 98
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 3 highly water endangering
Disposal 
6
Sulfides and sulfites as well as other reducing substances, inorganic cyanides and isonitriles: Stir into sodium hypochlorite solution (Cat. No. 105614) and, if necessary, leave to react for several days. Caution: some substances may show a violent reaction! Draw off any toxic or flammable gases that may be formed. Excess oxidant should be neutralized with sodium thiosulfate (Cat. No. 106513). Container D or E.
Declaration (railroad and road) ADR, RID 
Kein Gefahrgut
Declaration (transport by air) IATA-DGR 
UN 3334 , 9, III
Declaration (transport by sea) IMDG-Code 
No Dangerous Good
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?