• USA Home
  • 8.41195 - Phenylsulfonylacetone

EMAIL THIS PAGE TO A FRIEND
8.41195 EMD Millipore

Phenylsulfonylacetone

Phenylsulfonylacetone for synthesis. CAS 5000-44-2, molar mass 198.23 g/mol., for synthesis

Synonym: Phenylsulfonylpropan-2-one

  • CAS Number 5000-44-2

  • Linear Formula C₉H₁₀O₃S

  • Molecular Weight 198.23 g/mol

Purchase

Properties

Related Categories Biochemicals and Reagents, Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfones,
packaging   1 g in Plastic bottle (8411950001)
mp   58 - 59 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Melting range (lower value): ≥ 55 °C
  Melting range (upper value): ≤ 58 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2930 90 98
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?