• USA Home
  • 8.41196 - Ethyl 3-oxovalerate

EMAIL THIS PAGE TO A FRIEND
8.41196 EMD Millipore

Ethyl 3-oxovalerate

Ethyl 3-oxovalerate for synthesis. CAS 4949-44-4, molar mass 144.17 g/mol., for synthesis

Synonym: Ethyl propionylacetate, Propionylacetic acid ethyl ester

  • CAS Number 4949-44-4

  • Linear Formula C₇H₁₂O₃

  • Molecular Weight 144.17 g/mol

  •  EC Number 225-593-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 ml in Glass bottle (8411960005)
bp   190 - 192 °C
density   1.01 g/cm3 (20 °C)
flash point   77 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.003 - 1.009
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2918 30 00
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?