• USA Home
  • 8.41197 - 4-Hydroxy-3-methoxybenzyl alcohol

EMAIL THIS PAGE TO A FRIEND
8.41197 EMD Millipore

4-Hydroxy-3-methoxybenzyl alcohol

4-Hydroxy-3-methoxybenzyl alcohol for synthesis. CAS 498-00-0, molar mass 154.16 g/mol., for synthesis

Synonym: Vanillyl alcohol

  • CAS Number 498-00-0

  • Linear Formula C₈H₁₀O₃

  • Molecular Weight 154.16 g/mol

  •  EC Number 207-852-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   50 g in Plastic bottle (8411970050)
mp   113 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 112 °C
  Melting range (upper value): ≤ 115 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2909 50 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?