• USA Home
  • 8.41199 - Dimethyl acetylmethylphosphonate

EMAIL THIS PAGE TO A FRIEND
8.41199 EMD Millipore

Dimethyl acetylmethylphosphonate

Dimethyl acetylmethylphosphonate for synthesis. CAS 4202-14-6, molar mass 166.11 g/mol., for synthesis

Synonym: (2-Oxo-propyl)-phosphonic acid dimethyl ester

  • CAS Number 4202-14-6

  • Linear Formula C₅H₁₁O₄P

  • Molecular Weight 166.11 g/mol

  •  EC Number 224-110-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 ml in Glass bottle (8411990005)
bp   121 - 123 °C (14.7 hPa)
density   1.2 g/cm3 (20 °C)
flash point   93 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 95.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.205 - 1.210
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2931 39 90
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?