• USA Home
  • 8.41201 - 1,2-Diaminopropane-tetraacetic acid

EMAIL THIS PAGE TO A FRIEND
8.41201 EMD Millipore

1,2-Diaminopropane-tetraacetic acid

1,2-Diaminopropane-tetraacetic acid for synthesis. CAS 4408-81-5, molar mass 306.27 g/mol., for synthesis

  • CAS Number 4408-81-5

  • Linear Formula C₁₁H₁₈N₂O₈

  • Molecular Weight 306.27 g/mol

  •  EC Number 224-559-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Plastic bottle (8412010005)
mp   241 °C (decomposition)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (acidimetric): ≥ 98.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2922 49 85
Hazard Statement(s) 
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
R Phrase 
R 36
Irritating to eyes.
S Phrase 
S 26
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?