• USA Home
  • 8.41206 - 2'-Fluoroacetophenone

EMAIL THIS PAGE TO A FRIEND
8.41206 EMD Millipore

2'-Fluoroacetophenone

2'-Fluoroacetophenone for synthesis. CAS 445-27-2, molar mass 138.14 g/mol., for synthesis

Synonym: 1-(2-Fluorophenyl)ethanone

  • CAS Number 445-27-2

  • Linear Formula C₈H₇FO

  • Molecular Weight 138.14 g/mol

  •  EC Number 207-156-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 ml in Glass bottle (8412060005)
bp   187 - 189 °C (1013 hPa)
density   1.137 g/cm3 (20 °C)
flash point   61 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.136 - 1.140
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2914 79 00
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Categories of danger 
irritant
R Phrase 
R 36/38
Irritating to eyes and skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?