• USA Home
  • 8.41207 - 4-Bromobenzotrifluoride

EMAIL THIS PAGE TO A FRIEND
8.41207 EMD Millipore

4-Bromobenzotrifluoride

4-Bromobenzotrifluoride for synthesis. CAS 402-43-7, molar mass 225 g/mol., for synthesis

Synonym: p-Bromo-α,α,α-trifluorotoluene, 1-Bromo-4-(trifluoromethyl)benzene

  • CAS Number 402-43-7

  • Linear Formula C₇H₄BrF₃

  • Molecular Weight 225 g/mol

  •  EC Number 206-943-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 ml in Glass bottle (8412070005)
density   1.63 g/cm3 (20 °C)
flash point   48 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.628 - 1.632
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2903 99 80
Hazard Statement(s) 
H226: Flammable liquid and vapour.
H315: Causes skin irritation.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P210: Keep away from heat.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
3 Flammable liquids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Declaration (railroad and road) ADR, RID 
UN 1993 , 3, II
Declaration (transport by air) IATA-DGR 
UN 1993 , 3, II
Declaration (transport by sea) IMDG-Code 
UN 1993 , 3, II
Categories of danger 
flammable, irritant
R Phrase 
R 10-36/38
Flammable.Irritating to eyes and skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?