• USA Home
  • 8.41208 - 2-Fluorobenzoyl chloride

EMAIL THIS PAGE TO A FRIEND
8.41208 EMD Millipore

2-Fluorobenzoyl chloride

2-Fluorobenzoyl chloride for synthesis. CAS 393-52-2, molar mass 158.55 g/mol., for synthesis

Synonym: 2-Fluoro-benzoic acid chloride

  • CAS Number 393-52-2

  • Linear Formula C₇H₄ClFO

  • Molecular Weight 158.55 g/mol

  •  EC Number 206-887-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8412080025)
bp   89 - 91 °C (18.7 hPa)
density   1.331 g/cm3 (20 °C)
flash point   82 °C
mp   4 - 6 °C
LD 50 oral   LD50 Rat 2600 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.332 - 1.337
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2916 39 90
Hazard Statement(s) 
H314: Causes severe skin burns and eye damage.
Precautionary Statement(s) 
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
RTECS 
DM6640000
Storage class 
8A Combustible, corrosive hazardous materials
WGK 
WGK 1 slightly water endangering
Disposal 
11
Organic acid halides, anhydrides and isocyanates can be added dropwise to an excess of methanol (Cat. No. 822283) to convert them into the corresponding methyl esters or methyl carbamates. If necessary, neutralize with sodium hydroxide solution (Cat. No. 105587). Fill into container A.
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Symbols 
Corrosive  Corrosive
Declaration (railroad and road) ADR, RID 
UN 1760 , 8, II
Declaration (transport by air) IATA-DGR 
UN 1760 , 8, II
Declaration (transport by sea) IMDG-Code 
UN 1760 , 8, II
Categories of danger 
corrosive
R Phrase 
R 34
Causes burns.
S Phrase 
S 26-36/37/39-45
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing, gloves and eye/face protection.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?