• USA Home
  • 8.41210 - 4'-Fluoroacetophenone

EMAIL THIS PAGE TO A FRIEND
8.41210 EMD Millipore

4'-Fluoroacetophenone

4'-Fluoroacetophenone for synthesis. CAS 403-42-9, molar mass 138.14 g/mol., for synthesis

  • CAS Number 403-42-9

  • Linear Formula C₈H₇FO

  • Molecular Weight 138.14 g/mol

  •  EC Number 206-960-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8412100025)
density   1.141 g/cm3 (20 °C)
flash point   71 °C
mp   4 °C
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.140 - 1.144
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2914 79 00
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Categories of danger 
irritant
R Phrase 
R 36/38
Irritating to eyes and skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?