• USA Home
  • 8.41211 - 2-Fluorobenzonitrile

EMAIL THIS PAGE TO A FRIEND
8.41211 EMD Millipore

2-Fluorobenzonitrile

2-Fluorobenzonitrile for synthesis. CAS 394-47-8, molar mass 121.11 g/mol., for synthesis

  • CAS Number 394-47-8

  • Linear Formula C₇H₄FN

  • Molecular Weight 121.11 g/mol

  •  EC Number 206-897-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 ml in Glass bottle (8412110010)
bp   84 - 86 °C (18.7 hPa)
density   1.138 g/cm3 (20 °C)
flash point   74 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.136 - 1.140
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2926 90 70
Hazard Statement(s) 
H302 + H312: Harmful if swallowed or in contact with skin
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 3276 , 6.1, II
Declaration (transport by air) IATA-DGR 
UN 3276 , 6.1, II
Declaration (transport by sea) IMDG-Code 
UN 3276 , 6.1, II
Categories of danger 
harmful
R Phrase 
R 21/22
Harmful in contact with skin and if swallowed.
S Phrase 
S 36/37
Wear suitable protective clothing and gloves.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?