• USA Home
  • 8.41218 - 3-Cyanobenzaldehyde

EMAIL THIS PAGE TO A FRIEND
8.41218 EMD Millipore

3-Cyanobenzaldehyde

3-Cyanobenzaldehyde for synthesis. CAS 24964-64-5, molar mass 131.13 g/mol., for synthesis

Synonym: 3-Formylbenzonitrile

  • CAS Number 24964-64-5

  • Linear Formula C₈H₅NO

  • Molecular Weight 131.13 g/mol

  •  EC Number 246-549-1

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   1 g in Plastic bottle (8412180001)
bp   209 - 210 °C (1.013 hPa)
mp   79 - 80 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Melting range (lower value): ≥ 77 °C
  Melting range (upper value): ≤ 80 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2926 90 70
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
H335: May cause respiratory irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Declaration (railroad and road) ADR, RID 
UN 2811 , 6.1, II
Declaration (transport by air) IATA-DGR 
UN 2811 , 6.1, II
Declaration (transport by sea) IMDG-Code 
UN 2811 , 6.1, II
Categories of danger 
irritant
R Phrase 
R 36/37/38
Irritating to eyes, respiratory system and skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?