EMAIL THIS PAGE TO A FRIEND
8.41219 EMD Millipore

Epoxycyclododecane

Epoxycyclododecane for synthesis. CAS 286-99-7, molar mass 182.3 g/mol., for synthesis

Synonym: Cyclododeceneoxid

  • CAS Number 286-99-7

  • Linear Formula C₁₂H₂₂O

  • Molecular Weight 182.3 g/mol

  •  EC Number 206-012-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8412190025)
density   0.939 g/cm3 (20 °C)
flash point   >110 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay isomers (GC, area%): ≥ 95.0 % (a/a)
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2910 90 00
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?