• USA Home
  • 8.41221 - 2-(Trimethylsilyl)-ethanol

EMAIL THIS PAGE TO A FRIEND
8.41221 EMD Millipore

2-(Trimethylsilyl)-ethanol

2-(Trimethylsilyl)-ethanol for synthesis. CAS 2916-68-9, EC Number 220-844-5, molar mass 118.25 g/mol., for synthesis

Synonym: 2-Hydroxyethyltrimethyl silane

  • CAS Number 2916-68-9

  • Linear Formula C₅H₁₄OSi

  • Molecular Weight 118.25 g/mol

  •  EC Number 220-844-5

Purchase

Properties

packaging   5 ml in Glass bottle (8412210005)
bp   71 - 72 °C (46.7 hPa)
density   0.825 g/cm3 (20 °C)
flash point   50 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.828 - 0.832
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2931 90 00
Hazard Statement(s) 
H226: Flammable liquid and vapour.
Precautionary Statement(s) 
P210: Keep away from heat.
Storage class 
3 Flammable liquids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Declaration (railroad and road) ADR, RID 
UN 1993 , 3, III
Declaration (transport by air) IATA-DGR 
UN 1993 , 3, III
Declaration (transport by sea) IMDG-Code 
UN 1993 , 3, III
Categories of danger 
flammable
R Phrase 
R 10
Flammable.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?