EMAIL THIS PAGE TO A FRIEND
8.41222 EMD Millipore

3-Chloroanisole

3-Chloroanisole for synthesis. CAS 2845-89-8, molar mass 142.58 g/mol., for synthesis

Synonym: 1-Chloro-3-methoxybenzene

  • CAS Number 2845-89-8

  • Linear Formula C₇H₇ClO

  • Molecular Weight 142.58 g/mol

  •  EC Number 220-642-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   50 ml in Glass bottle (8412220050)
bp   193 °C (1013 hPa)
solubility   235 mg/l
density   1.172 g/cm3 (20 °C)
flash point   73 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.170 - 1.174
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2909 30 90
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?