• USA Home
  • 8.41223 - 2,6-Dimethylcyclohexanone

EMAIL THIS PAGE TO A FRIEND
8.41223 EMD Millipore

2,6-Dimethylcyclohexanone

2,6-Dimethylcyclohexanone (mixture of cis and trans isomers) for synthesis. CAS 2816-57-1, molar mass 126.19 g/mol., (mixture of cis and trans isomers) for synthesis

  • CAS Number 2816-57-1

  • Linear Formula C₈H₁₄O

  • Molecular Weight 126.19 g/mol

  •  EC Number 220-570-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8412230025)
bp   174 - 176 °C (1013 hPa)
density   0.92 g/cm3 (20 °C)
flash point   51 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.910 - 0.914
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2914 29 00
Hazard Statement(s) 
H226: Flammable liquid and vapour.
Precautionary Statement(s) 
P210: Keep away from heat.
Storage class 
3 Flammable liquids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Declaration (railroad and road) ADR, RID 
UN 1224 , 3, III
Declaration (transport by air) IATA-DGR 
UN 1224 , 3, III
Declaration (transport by sea) IMDG-Code 
UN 1224 , 3, III
Categories of danger 
flammable
R Phrase 
R 10
Flammable.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?