EMAIL THIS PAGE TO A FRIEND
8.41229 EMD Millipore

Indazole

Indazole for synthesis. CAS 271-44-3, molar mass 118.14 g/mol., for synthesis

Synonym: Benzpyrazole, Isoindazone

  • CAS Number 271-44-3

  • Linear Formula C₇H₆N₂

  • Molecular Weight 118.14 g/mol

  •  EC Number 205-978-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   1 g in Plastic bottle (8412290001)
mp   147 - 149 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 145 °C
  Melting range (upper value): ≤ 148 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 99 80
RTECS 
NK7745000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?