• USA Home
  • 8.41230 - 4-Bromo-1,2-dimethoxybenzene

EMAIL THIS PAGE TO A FRIEND
8.41230 EMD Millipore

4-Bromo-1,2-dimethoxybenzene

4-Bromo-1,2-dimethoxybenzene for synthesis. CAS 2859-78-1, molar mass 217.06 g/mol., for synthesis

Synonym: 4-Bromoveratrole

  • CAS Number 2859-78-1

  • Linear Formula C₈H₉BrO₂

  • Molecular Weight 217.06 g/mol

  •  EC Number 220-677-8

Purchase

Properties

packaging   25 ml in Glass bottle (8412300025)
bp   124 - 126 °C (12 hPa)
density   1.51 g/cm3 (20 °C)
flash point   109 °C
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.500 - 1.510
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2909 30 38
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?