EMAIL THIS PAGE TO A FRIEND
8.41237 EMD Millipore

Thiazole

Thiazole for synthesis. CAS 288-47-1, molar mass 85.12 g/mol., for synthesis

  • CAS Number 288-47-1

  • Linear Formula C₃H₃NS

  • Molecular Weight 85.12 g/mol

  •  EC Number 206-021-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 ml in Glass bottle (8412370005)
bp   117 - 118 °C (1013 hPa)
density   1.2 g/cm3 (20 °C)
flash point   22 °C
mp   -33 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.197 - 1.201
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2934 10 00
Hazard Statement(s) 
H225: Highly flammable liquid and vapour.
H302: Harmful if swallowed.
Precautionary Statement(s) 
P210: Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
P240: Ground/bond container and receiving equipment.
P403 + P233: Store in a well-ventilated place. Keep container tightly closed.
Storage class 
3 Flammable liquids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 1993 , 3, II
Declaration (transport by air) IATA-DGR 
UN 1993 , 3, II
Declaration (transport by sea) IMDG-Code 
UN 1993 , 3, II
Categories of danger 
flammable
R Phrase 
R 10-22
Flammable.Harmful if swallowed.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?