• USA Home
  • 8.41238 - Biphenyl-4-carbaldehyde

EMAIL THIS PAGE TO A FRIEND
8.41238 EMD Millipore

Biphenyl-4-carbaldehyde

Biphenyl-4-carbaldehyde for synthesis. CAS 3218-36-8, molar mass 182.22 g/mol., for synthesis

Synonym: 4-Phenylbenzaldehyde

  • CAS Number 3218-36-8

  • Linear Formula C₁₃H₁₀O

  • Molecular Weight 182.22 g/mol

  •  EC Number 221-742-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Glass bottle (8412380010)
bp   184 °C (14.7 hPa)
mp   58 - 60 °C
shipped in   ambient
storage temp.   Store at -20°C to 0°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 56 °C
  Melting range (upper value): ≤ 59 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2912 29 00
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Declaration (railroad and road) ADR, RID 
Kein Gefahrgut
Declaration (transport by air) IATA-DGR 
No Dangerous Good
Declaration (transport by sea) IMDG-Code 
No Dangerous Good
Categories of danger 
irritant
R Phrase 
R 36/38
Irritating to eyes and skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?