EMAIL THIS PAGE TO A FRIEND
8.41245 EMD Millipore

Cyclopentadecanone

Cyclopentadecanone for synthesis. CAS 502-72-7, molar mass 224.38 g/mol., for synthesis

Synonym: Exaltone

  • CAS Number 502-72-7

  • Linear Formula C₁₅H₂₈O

  • Molecular Weight 224.38 g/mol

  •  EC Number 207-951-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Plastic bottle (8412450005)
bp   120 °C (0.4 hPa)
density   0.897 g/cm3 (25 °C)
mp   64 - 66 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 63 °C
  Melting range (upper value): ≤ 66 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2914 29 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?