EMAIL THIS PAGE TO A FRIEND
8.41253 EMD Millipore

4-Allylanisole

4-Allylanisole for synthesis. CAS 140-67-0, molar mass 148.2 g/mol., for synthesis

Synonym: Estragole, 4-Methoxyallylbenzene

  • CAS Number 140-67-0

  • Linear Formula C₁₀H₁₂O

  • Molecular Weight 148.2 g/mol

  •  EC Number 205-427-8

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8412530025)
solubility   0.178 g/l
density   0.965 g/cm3 (20 °C)
flash point   81 °C
LD 50 oral   LD50 Rat 1230 mg/kg
LD 50 dermal   LD50 Rabbit > 5000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.965 - 0.970
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2909 30 90
Hazard Statement(s) 
H302: Harmful if swallowed.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Categories of danger 
harmful
R Phrase 
R 22
Harmful if swallowed.
S Phrase 
S 24/25
Avoid contact with skin and eyes.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?