• USA Home
  • 8.41258 - Di-tert-butyl malonate

EMAIL THIS PAGE TO A FRIEND
8.41258 EMD Millipore

Di-tert-butyl malonate

Di-tert-butyl malonate for synthesis. CAS 541-16-2, molar mass 216.27 g/mol., for synthesis

Synonym: Malonic acid-di-tert-butylester

  • CAS Number 541-16-2

  • Linear Formula C₁₁H₂₀O₄

  • Molecular Weight 216.27 g/mol

  •  EC Number 208-769-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 ml in Glass bottle (8412580010)
bp   65 - 67 °C (1.33 hPa)
density   0.965 g/cm3 (20 °C)
flash point   88 °C
mp   -7 °C
shipped in   ambient
storage temp.   Store at <= 20°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.961 - 0.966
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2917 19 10
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?